2-Amino-3,5-difluorophenol 99%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,4-Difluoro-6-hydroxyaniline
PC8303-1G
PC8303-5G
APOSPC8303-1GEA
4570
CZK
APOSPC8303-1G
APOSPC8303-5G
2-Amino-3,5-difluorophenol 99%
2-Amino-3,5-difluorophenol
Vzorec:
C₆H₅F₂NO MW: 145,107 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03094490 CAS číslo: 163733-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...