Tisk…

1-Iodo-1H,1H,2H,2H-perfluoro(4,7,10,13,16-pentamethyl-5,8,11,14,17-pentaoxaeicosane) 95%

Dodavatel: Apollo Scientific

PC8266-1G PC8266-5G
APOSPC8266-1GEA 2570 CZK
APOSPC8266-1G APOSPC8266-5G
1-Iodo-1H,1H,2H,2H-perfluoro(4,7,10,13,16-pentamethyl-5,8,11,14,17-pentaoxaeicosane) 95%
1-Iodo-1H,1H,2H,2H-perfluoro(4,7,10,13,16-pentamethyl-5,8,11,14,17-pentaoxaeicosane)
Vzorec: C₂₀H₄F₃₇IO₅
MW: 1154,088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460486
CAS číslo: 1212369-20-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR