Perfluoro(1-iodo-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecane) 95+%
Dodavatel: Apollo Scientific
|
Varování
|
PC8265-1G
PC8265-5G
APOSPC8265-1GEA
2330
CZK
APOSPC8265-1G
APOSPC8265-5G
Perfluoro(1-iodo-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecane) 95+%
Perfluoro(1-iodo-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecane)
Vzorec:
C₁₈F₃₇IO₅ MW: 1126,035 g/mol Bod varu: 90 @ 0,4mm °C Storage Temperature: Ambient |
MDL Number:
MFCD08460485 CAS číslo: 1212223-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...