1H,1H,2H,2H-Perfluoro(8,8-dimethyl-1-iodo-7-oxaoctane)
Dodavatel: Apollo Scientific
Synonyms:
6-(Heptafluoroisopropoxy)-1-iodo-1H,1H,2H,2H-octafluorohexane
PC8262-1G
PC8262-5G
APOSPC8262-1GEA
2040
CZK
APOSPC8262-1G
APOSPC8262-5G
1H,1H,2H,2H-Perfluoro(8,8-dimethyl-1-iodo-7-oxaoctane)
1H,1H,2H,2H-Perfluoro(8,8-dimethyl-1-iodo-7-oxaoctane)
Vzorec:
C₉H₄F₁₅IO Storage Temperature: Ambient |
MDL Number:
MFCD09763608 CAS číslo: 25080-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...