4-Bromo-2,3,6-trifluorobenzyl alcohol 95%
Dodavatel: Apollo Scientific
PC8248-1G
PC8248-250MG
APOSPC8248-1GEA
17990
CZK
APOSPC8248-1G
APOSPC8248-250MG
4-Bromo-2,3,6-trifluorobenzyl alcohol 95%
4-Bromo-2,3,6-trifluorobenzyl alcohol
Vzorec:
C₇H₄BrF₃O MW: 241,005 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094469 CAS číslo: 252004-34-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...