Tisk…

4-Bromo-2,3,6-trifluorobenzaldehyde 95%

Dodavatel: Apollo Scientific

PC8247-1G PC8247-250MG PC8247-5G
APOSPC8247-1GEA 3020 CZK
APOSPC8247-1G APOSPC8247-250MG APOSPC8247-5G
4-Bromo-2,3,6-trifluorobenzaldehyde 95%
4-Bromo-2,3,6-trifluorobenzaldehyde
Vzorec: C₇H₂BrF₃O
MW: 238,989 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094468
CAS číslo: 537033-56-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR