5-(4-Bromophenylsulphonyl)-4-(hydroxymethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Dodavatel: Apollo Scientific
Synonyms:
[5-(4-Bromophenyl)sulphonyl-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
PC8127-1G
PC8127-500MG
APOSPC8127-1GEA
33400
CZK
APOSPC8127-1G
APOSPC8127-500MG
5-(4-Bromophenylsulphonyl)-4-(hydroxymethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
5-(4-Bromophenylsulphonyl)-4-(hydroxymethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Vzorec:
C₁₂H₁₀BrF₃N₂O₃S MW: 399,184 g/mol Bod tání: 124…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00172661 CAS číslo: 318469-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...