Tisk…

5-(4-Bromophenylthio)-4-(hydroxymethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole 97%

Dodavatel: Apollo Scientific

Synonyms: [5-(4-Bromophenylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol

PC8124-1G PC8124-500MG
APOSPC8124-1GEA 22390 CZK
APOSPC8124-1G APOSPC8124-500MG
5-(4-Bromophenylthio)-4-(hydroxymethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole 97%
5-(4-Bromophenylthio)-4-(hydroxymethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Vzorec: C₁₂H₁₀BrF₃N₂OS
MW: 367,185 g/mol
Bod tání: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00172639
CAS číslo: 318239-56-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR