2,6-Dimethyl-4-fluorobenzaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Fluoro-3-formyl-m-xylene, 4-Fluoro-2,6-dimethylbenzaldehyde
PC8085-1G
PC8085-250MG
PC8085-5G
APOSPC8085-1GEA
9890
CZK
APOSPC8085-1G
APOSPC8085-250MG
APOSPC8085-5G
2,6-Dimethyl-4-fluorobenzaldehyde
2,6-Dimethyl-4-fluorobenzaldehyde
Vzorec:
C₉H₉FO MW: 152.166 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03701054 CAS číslo: 925441-35-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...