Nonafluoropentane 85%
Dodavatel: Apollo Scientific
|
Varování
|
PC8067-1G
PC8067-5G
APOSPC8067-1GEA
2490
CZK
APOSPC8067-1G
APOSPC8067-5G
Nonafluoropentane 85%
Nonafluoropentane
Mixture of 2H,3H,3H- and 2H,2H,3H- isomers
Vzorec:
C₅H₃F₉ MW: 234,063 g/mol Bod varu: 49…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD09258989 CAS číslo: 141993-31-9 UN: 1993 ADR: 3,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...