4-Fluoro-3-iodo-1-methyl-1H-indazole
Dodavatel: Apollo Scientific
|
Varování
|
PC8066-1G
PC8066-250MG
PC8066-5G
APOSPC8066-1GEA
9530
CZK
APOSPC8066-1G
APOSPC8066-250MG
APOSPC8066-5G
4-Fluoro-3-iodo-1-methyl-1H-indazole
4-Fluoro-3-iodo-1-methyl-1H-indazole
Vzorec:
C₈H₆FIN₂ MW: 276.05 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12756570 CAS číslo: 1257535-07-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...