Tisk…

6-Fluoro-3-iodo-1-methyl-1H-indazole 95%

Dodavatel: Apollo Scientific
Varování

PC7949-1G PC7949-250MG PC7949-100MG
APOSPC7949-1GEA 9460 CZK
APOSPC7949-1G APOSPC7949-250MG APOSPC7949-100MG
6-Fluoro-3-iodo-1-methyl-1H-indazole 95%
6-Fluoro-3-iodo-1-methyl-1H-indazole
Vzorec: C₈H₆FIN₂
MW: 276,05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12756569
CAS číslo: 1257535-15-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR