Tisk…

2-Bromo-3,3,3-trifluoropropan-1-ol 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-2-hydroxy-1-(trifluoromethyl)ethane, 2-Bromo-3-hydroxy-1,1,1-trifluoropropane

PC7935-5G
APOSPC7935-5GEA 2280 CZK
APOSPC7935-5G
2-Bromo-3,3,3-trifluoropropan-1-ol 95%
2-Bromo-3,3,3-trifluoropropan-1-ol
Vzorec: C₃H₄BrF₃O
MW: 192,962 g/mol
Bod varu: 34 °C (12 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09763638
CAS číslo: 311-86-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR