1H,1H,2H,2H,3H,3H,4H,4H-Perfluorodecan-1-ol
Dodavatel: Apollo Scientific
|
Varování
|
PC7931-1G
PC7931-250MG
PC7931-5G
APOSPC7931-1GEA
13330
CZK
APOSPC7931-1G
APOSPC7931-250MG
APOSPC7931-5G
1H,1H,2H,2H,3H,3H,4H,4H-Perfluorodecan-1-ol
1H,1H,2H,2H,3H,3H,4H,4H-Perfluorodecan-1-ol
Vzorec:
C₁₀H₉F₁₃O MW: 392,157 g/mol Bod varu: 80 °C (7 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD09864675 CAS číslo: 39239-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...