1-Chloro-1,2,3,3,3-pentafluoroprop-1-ene
Dodavatel: Apollo Scientific
PC7929-25G
PC7929-5G
APOSPC7929-25GEA
11610
CZK
APOSPC7929-25G
APOSPC7929-5G
1-Chloro-1,2,3,3,3-pentafluoroprop-1-ene
1-Chloro-1,2,3,3,3-pentafluoroprop-1-ene
Vzorec:
C₃ClF₅ MW: 166,477 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09998196 CAS číslo: 2804-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...