Tisk…

2-Bromo-5-(trifluoromethyl)benzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Bromo-3-(bromomethyl)benzotrifluoride, 1-Bromo-2-(bromomethyl)-4-(trifluoromethyl)benzene

PC7887-1G PC7887-5G PC7887-10G
APOSPC7887-1GEA 953 CZK
APOSPC7887-1G APOSPC7887-5G APOSPC7887-10G
2-Bromo-5-(trifluoromethyl)benzyl bromide 97%
2-Bromo-5-(trifluoromethyl)benzyl bromide
Vzorec: C₈H₅Br₂F₃
MW: 317,93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973762
CAS číslo: 886496-63-5
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR