Ethyl 4-chloro-2-fluorobenzoate
Dodavatel: Apollo Scientific
|
Varování
|
PC7776-25G
PC7776-5G
APOSPC7776-25GEA
5460
CZK
APOSPC7776-25G
APOSPC7776-5G
Ethyl 4-chloro-2-fluorobenzoate
Ethyl 4-chloro-2-fluorobenzoate
Vzorec:
C₉H₈ClFO₂ MW: 202,61 g/mol Bod varu: 206 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095401 CAS číslo: 4793-20-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...