2-Chloro-2,3,3-trifluorocyclobutane-1-carbonitrile 97+%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC7748-10G
PC7748-1G
PC7748-5G
APOSPC7748-10GEA
19680
CZK
APOSPC7748-10G
APOSPC7748-1G
APOSPC7748-5G
2-Chloro-2,3,3-trifluorocyclobutane-1-carbonitrile 97+%
2-Chloro-2,3,3-trifluorocyclobutane-1-carbonitrile
Vzorec:
C₅H₃ClF₃N Bod varu: 82 °C (40 mm Hg) Bod vzplanutí: 72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039436 CAS číslo: 1546-77-6 UN: 1992 ADR: 3+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...