2-Bromo-5-fluorobenzoyl chloride, tech
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC7745-1G
PC7745-5G
APOSPC7745-1GEA
3740
CZK
APOSPC7745-1G
APOSPC7745-5G
2-Bromo-5-fluorobenzoyl chloride, tech
2-Bromo-5-fluorobenzoyl chloride
Vzorec:
C₇H₃BrClFO MW: 237,454 g/mol Bod tání: 20…22 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094438 CAS číslo: 111771-13-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...