4-[3,5-Bis(trifluoromethyl)phenyl]iodobenzene
Dodavatel: Apollo Scientific
|
Varování
|
PC7691-1G
PC7691-5G
APOSPC7691-1GEA
3970
CZK
APOSPC7691-1G
APOSPC7691-5G
4-[3,5-Bis(trifluoromethyl)phenyl]iodobenzene
4-[3,5-Bis(trifluoromethyl)phenyl]iodobenzene
Vzorec:
C₁₄H₇F₆I MW: 416,1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368762 CAS číslo: 886762-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...