2,4-Difluoro-3-(trifluoromethyl)phenacyl bromide 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Bromo-1-[2,4-difluoro-3-(trifluoromethyl)phenyl]ethan-1-one, 2-Bromo-2',4'-difluoro-3'-(trifluoromethyl)acetophenone
PC7675-1G
PC7675-250MG
APOSPC7675-1GEA
27440
CZK
APOSPC7675-1G
APOSPC7675-250MG
2,4-Difluoro-3-(trifluoromethyl)phenacyl bromide 95%
2,4-Difluoro-3-(trifluoromethyl)phenacyl bromide
Vzorec:
C₉H₄BrF₅O MW: 303.023 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12547947 CAS číslo: 1211707-65-1 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...