Tisk…

4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl bromide 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-(Bromomethyl)-5-(methylsulphonyl)benzotrifluoride, 4-(Bromomethyl)-3-(trifluoromethyl)phenyl methyl sulphone, 1-(Bromomethyl)-4-(methylsulphonyl)-2-(trifluoromethyl)benzene

PC7521-1G PC7521-5G PC7521-10G
APOSPC7521-1GEA 8270 CZK
APOSPC7521-1G APOSPC7521-5G APOSPC7521-10G
4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl bromide 98%
4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl bromide
Vzorec: C₉H₈BrF₃O₂S
MW: 317.123 g/mol
Bod tání: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD16606069
CAS číslo: 934557-65-0
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR