Tisk…
2,4-Difluorophenyl trifluoromethanesulphonate 97%
Dodavatel: Apollo Scientific
Synonyms: 2,4-Difluorophenyl triflate
PC7501-1G PC7501-5G PC7501-250MG
APOSPC7501-1GEA 5630 CZK
APOSPC7501-1G APOSPC7501-5G APOSPC7501-250MG
2,4-Difluorophenyl trifluoromethanesulphonate 97%
2,4-Difluorophenyl trifluoromethanesulphonate
Vzorec: C₇H₃F₅O₃S
MW: 262.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784321
CAS číslo: 264135-49-1