Tisk…

2-Amino-3-bromo-5-(trifluoromethoxy)benzoic acid

Dodavatel: Apollo Scientific

PC7419-1G PC7419-5G
APOSPC7419-1GEA 2780 CZK
APOSPC7419-1G APOSPC7419-5G
2-Amino-3-bromo-5-(trifluoromethoxy)benzoic acid
2-Amino-3-bromo-5-(trifluoromethoxy)benzoic acid
Vzorec: C₈H₅BrF₃NO₃
MW: 300,029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407438
CAS číslo: 874774-41-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR