Tisk…

2-(4-Bromo-2-fluorophenoxy)propane

Dodavatel: Apollo Scientific

PC7409-1G PC7409-5G PC7409-10G
APOSPC7409-1GEA 1040 CZK
APOSPC7409-1G APOSPC7409-5G APOSPC7409-10G
2-(4-Bromo-2-fluorophenoxy)propane
2-(4-Bromo-2-fluorophenoxy)propane
Vzorec: C₉H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD00070760
CAS číslo: 202865-80-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR