3-Bromo-4-fluorobenzaldoxime
Dodavatel: Apollo Scientific
|
Varování
|
PC7367-1G
PC7367-5G
APOSPC7367-1GEA
2650
CZK
APOSPC7367-1G
APOSPC7367-5G
3-Bromo-4-fluorobenzaldoxime
3-Bromo-4-fluorobenzaldoxime
Vzorec:
C₇H₅BrFNO MW: 218.023 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00143092 CAS číslo: 202865-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...