Tisk…

5,5,5-Trifluoropentanonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5,5,5-Trifluorovaleronitrile

PC7318-1G PC7318-5G PC7318-10G
APOSPC7318-1GEA 1870 CZK
APOSPC7318-1G APOSPC7318-5G APOSPC7318-10G
5,5,5-Trifluoropentanonitrile 98%
5,5,5-Trifluoropentanonitrile
Vzorec: C₅H₆F₃N
MW: 137.103 g/mol
Bod varu: 60 °C (10 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08059547
CAS číslo: 89866-61-5
UN: 3275
ADR: 6.1+3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR