[2-(Trifluoromethoxy)phenyl]methanesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-(Trifluoromethoxy)benzylsulphonyl chloride, [2-(trifluoromethoxy)phenyl]methanesulphonyl chloride
PC7251-1G
PC7251-250MG
APOSPC7251-1GEA
7440
CZK
APOSPC7251-1G
APOSPC7251-250MG
[2-(Trifluoromethoxy)phenyl]methanesulphonyl chloride
[2-(Trifluoromethoxy)phenyl]methanesulphonyl chloride
Vzorec:
C₈H₆ClF₃O₃S MW: 274,645 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09749658 CAS číslo: 116827-38-4 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...