Tisk…

DL-2-(Trifluoromethyl)norvaline

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-2-(trifluoromethyl)pentanoic acid

PC7241-100MG PC7241-50MG
APOSPC7241-100MGEA 6650 CZK
APOSPC7241-100MG APOSPC7241-50MG
DL-2-(Trifluoromethyl)norvaline
DL-2-(Trifluoromethyl)norvaline
Vzorec: C₆H₁₀F₃NO₂
MW: 185,144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320781
CAS číslo: 119305-52-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR