Tisk…

2,5-Difluoro-DL-phenylglycine

Dodavatel: Apollo Scientific
Varování

PC7231-1G PC7231-5G PC7231-10G
APOSPC7231-1GEA 9590 CZK
APOSPC7231-1G APOSPC7231-5G APOSPC7231-10G
2,5-Difluoro-DL-phenylglycine
2,5-Difluoro-DL-phenylglycine
Vzorec: C₈H₇F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD02662504
CAS číslo: 243448-17-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR