Tisk…

4-Amino-6-fluoro-2-methylquinoline

Dodavatel: Apollo Scientific

Synonyms: 6-Fluoro-2-methylquinolin-4-amine

PC7197-1G PC7197-500MG PC7197-250MG
APOSPC7197-1GEA 14190 CZK
APOSPC7197-1G APOSPC7197-500MG APOSPC7197-250MG
4-Amino-6-fluoro-2-methylquinoline
4-Amino-6-fluoro-2-methylquinoline
Vzorec: C₁₀H₉FN₂
MW: 176.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06254679
CAS číslo: 288151-49-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR