Tisk…

4-Hydrazino-7-(trifluoromethyl)quinoline

Dodavatel: Apollo Scientific

PC7187-1G PC7187-500MG
APOSPC7187-1GEA 10350 CZK
APOSPC7187-1G APOSPC7187-500MG
4-Hydrazino-7-(trifluoromethyl)quinoline
4-Hydrazino-7-(trifluoromethyl)quinoline
Vzorec: C₁₀H₈F₃N₃
MW: 227,186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272471
CAS číslo: 49612-01-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR