Tisk…

2-Bromo-5-chloro-4-fluoroaniline

Dodavatel: Apollo Scientific
Varování

PC7174-1G PC7174-25G PC7174-5G
APOSPC7174-1GEA 622 CZK
APOSPC7174-1G APOSPC7174-25G APOSPC7174-5G
2-Bromo-5-chloro-4-fluoroaniline
2-Bromo-5-chloro-4-fluoroaniline
Vzorec: C₆H₄BrClFN
Storage Temperature: Ambient
MDL Number: MFCD10698796
CAS číslo: 85462-59-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR