Tisk…

2-(4-Chlorophenyl)-4-(trifluoromethyl)-1H-imidazole

Dodavatel: Apollo Scientific

PC7158-1G PC7158-5G
APOSPC7158-1GEA 3740 CZK
APOSPC7158-1G APOSPC7158-5G
2-(4-Chlorophenyl)-4-(trifluoromethyl)-1H-imidazole
2-(4-Chlorophenyl)-4-(trifluoromethyl)-1H-imidazole
Vzorec: C₁₀H₆ClF₃N₂
MW: 246,616 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460500
CAS číslo: 33469-15-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR