Tisk…

4-(Pentafluorobenzyloxy)benzyl alcohol

Dodavatel: Apollo Scientific

PC7157-1G PC7157-5G
APOSPC7157-1GEA 6750 CZK
APOSPC7157-1G APOSPC7157-5G
4-(Pentafluorobenzyloxy)benzyl alcohol
4-(Pentafluorobenzyloxy)benzyl alcohol
Vzorec: C₁₄H₉F₅O₂
MW: 304.212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09749654
CAS číslo: 471469-89-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR