Tisk…

(2Z)-4-Bromo-1,1,1,3-tetrafluorobut-2-ene 97%

Dodavatel: Apollo Scientific
Varování

PC7154-1G PC7154-5G
APOSPC7154-1GEA 3940 CZK
APOSPC7154-1G APOSPC7154-5G
(2Z)-4-Bromo-1,1,1,3-tetrafluorobut-2-ene 97%
(2Z)-4-Bromo-1,1,1,3-tetrafluorobut-2-ene
Vzorec: C₄H₃BrF₄
MW: 206,964 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460476
CAS číslo: 933600-89-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR