Tisk…

2-Bromo-5-(trifluoromethyl)phenylhydrazine hydrochloride

Dodavatel: Apollo Scientific
Varování

PC7112-1G PC7112-5G PC7112-25G
APOSPC7112-1GEA 992 CZK
APOSPC7112-1G APOSPC7112-5G APOSPC7112-25G
2-Bromo-5-(trifluoromethyl)phenylhydrazine hydrochloride
2-Bromo-5-(trifluoromethyl)phenylhydrazine hydrochloride
Vzorec: C₇H₆BrF₃N₂·ClH
MW: 291,496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094412
CAS číslo: 529512-78-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR