4-[5-(Trifluoromethyl)pyridin-2-yl]benzonitrile 97%
Dodavatel: Apollo Scientific
PC7105-1G
PC7105-5G
APOSPC7105-1GEA
4170
CZK
APOSPC7105-1G
APOSPC7105-5G
4-[5-(Trifluoromethyl)pyridin-2-yl]benzonitrile 97%
4-[5-(Trifluoromethyl)pyridin-2-yl]benzonitrile
Vzorec:
C₁₃H₇F₃N₂ MW: 248.203 g/mol Bod tání: 122.5…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD09064969 CAS číslo: 892501-99-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...