Tisk…

2-Morpholin-4-yl-5-(trifluoromethyl)benzeneboronic acid, pinacol ester 97%

Dodavatel: Apollo Scientific

PC7101-1G PC7101-250MG
APOSPC7101-1GEA 5720 CZK
APOSPC7101-1G APOSPC7101-250MG
2-Morpholin-4-yl-5-(trifluoromethyl)benzeneboronic acid, pinacol ester 97%
2-Morpholin-4-yl-5-(trifluoromethyl)benzeneboronic acid, pinacol ester
Vzorec: C₁₇H₂₃BF₃NO₃
Bod tání: 118…119 °C
Storage Temperature: Ambient
MDL Number: MFCD09064984
CAS číslo: 906352-77-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR