Tisk…

2-(Aminomethyl)-3-chloro-5-(trifluoromethyl)pyridine hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(Aminomethyl)-3-chloro-5-(trifluoromethyl)pyridine Hydrochloride, 3-Chloro-5-(trifluoromethyl)picolinylamine hydrochloride, [3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methylamine hydrochloride

PC7098-1G PC7098-250MG
APOSPC7098-1GEA 1490 CZK
APOSPC7098-1G APOSPC7098-250MG
2-(Aminomethyl)-3-chloro-5-(trifluoromethyl)pyridine hydrochloride
2-(Aminomethyl)-3-chloro-5-(trifluoromethyl)pyridine hydrochloride
Vzorec: C₇H₆ClF₃N₂·ClH
MW: 247,045 g/mol
Bod tání: 226…229 °C
Storage Temperature: Ambient
MDL Number: MFCD00210134
CAS číslo: 326476-49-7
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR