Tisk…

2-Bromo-4-fluorostyrene

Dodavatel: Apollo Scientific
Varování

PC7068-1G PC7068-5G
APOSPC7068-1GEA 3740 CZK
APOSPC7068-1G APOSPC7068-5G
2-Bromo-4-fluorostyrene
2-Bromo-4-fluorostyrene
Vzorec: C₈H₆BrF
Storage Temperature: Refrigerator
MDL Number: MFCD14892603
CAS číslo: 1221684-51-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR