1-(Chlorodifluoromethyl)-2-fluorobenzene
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
α-Chloro-α,α,2-trifluorotoluene
PC7047-1G
PC7047-5G
APOSPC7047-1GEA
8900
CZK
APOSPC7047-1G
APOSPC7047-5G
1-(Chlorodifluoromethyl)-2-fluorobenzene
1-(Chlorodifluoromethyl)-2-fluorobenzene
Vzorec:
C₇H₄ClF₃ MW: 180,555 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14892602 CAS číslo: 17054-13-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...