Tisk…

5-Bromo-2-fluorobenzeneboronic acid pinacol ester

Dodavatel: Apollo Scientific

Synonyms: 5-Bromo-2-fluorophenylboronic acid pinacol ester

PC6981-5G PC6981-25G
APOSPC6981-5GEA 3740 CZK
APOSPC6981-5G APOSPC6981-25G
5-Bromo-2-fluorobenzeneboronic acid pinacol ester
5-Bromo-2-fluorobenzeneboronic acid pinacol ester
Vzorec: C₁₂H₁₅BBrFO₂
MW: 300,96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09260441
CAS číslo: 942069-51-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR