3-(2,5-Difluorophenyl)propanoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC6962-25G
PC6962-5G
PC6962-1G
APOSPC6962-25GEA
7610
CZK
APOSPC6962-25G
APOSPC6962-5G
APOSPC6962-1G
3-(2,5-Difluorophenyl)propanoic acid 98%
3-(2,5-Difluorophenyl)propanoic acid
Vzorec:
C₉H₈F₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD04116053 CAS číslo: 130408-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...