4-Amino-2-fluoro-5-nitrobenzotrifluoride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
5-Fluoro-2-nitro-4-(trifluoromethyl)aniline, 2-Amino-4-fluoro-5-(trifluoromethyl)nitrobenzene
PC6958-1G
PC6958-250MG
APOSPC6958-1GEA
4140
CZK
APOSPC6958-1G
APOSPC6958-250MG
4-Amino-2-fluoro-5-nitrobenzotrifluoride
4-Amino-2-fluoro-5-nitrobenzotrifluoride
Vzorec:
C₇H₄F₄N₂O₂ Bod tání: 137…140 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09907681 CAS číslo: 428871-73-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...