Perfluoro-2,5-dimethyl-3,6-dioxanonanamide
Dodavatel: Apollo Scientific
Synonyms:
2-[1,1,2,3,3,3-Hexafluoro-2-(heptafluoropropoxy)propoxy]-2,3,3,3-tetrafluoropropanamide
PC6940-1G
PC6940-5G
APOSPC6940-1GEA
2670
CZK
APOSPC6940-1G
APOSPC6940-5G
Perfluoro-2,5-dimethyl-3,6-dioxanonanamide
Perfluoro-2,5-dimethyl-3,6-dioxanonanamide
Vzorec:
C₉H₂F₁₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00347264 CAS číslo: 60308-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...