5-Bromo-2-(trifluoromethoxy)benzonitrile 99+%
Dodavatel: Apollo Scientific
|
Varování
|
PC6919-1G
PC6919-5G
PC6919-10G
APOSPC6919-1GEA
622
CZK
APOSPC6919-1G
APOSPC6919-5G
APOSPC6919-10G
5-Bromo-2-(trifluoromethoxy)benzonitrile 99+%
5-Bromo-2-(trifluoromethoxy)benzonitrile
Vzorec:
C₈H₃BrF₃NO Storage Temperature: Ambient |
MDL Number:
MFCD13185875 CAS číslo: 1210906-15-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...