2,3-Difluoro-4-methylphenacyl bromide
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Bromo-2',3'-difluoro-4'-methylacetophenone, 2-Bromo-1-(2,3-difluoro-4-methylphenyl)ethan-1-one
PC6884-1G
PC6884-250MG
APOSPC6884-1GEA
29760
CZK
APOSPC6884-1G
APOSPC6884-250MG
2,3-Difluoro-4-methylphenacyl bromide
2,3-Difluoro-4-methylphenacyl bromide
Vzorec:
C₉H₇BrF₂O MW: 249.052 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD14581695 CAS číslo: 1221684-44-1 UN: 1759 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...