7-Iodoperfluoro-3-oxaheptanesulphonyl fluoride, tech
Dodavatel: Apollo Scientific
Synonyms:
2-(4-Iodo-1,1,2,2,3,3,4,4-octafluorobutoxy)-1,1,2,2-tetrafluoroethanesulphonyl fluoride
PC6876-1G
PC6876-250MG
PC6876-5G
APOSPC6876-1GEA
8010
CZK
APOSPC6876-1G
APOSPC6876-250MG
APOSPC6876-5G
7-Iodoperfluoro-3-oxaheptanesulphonyl fluoride, tech
7-Iodoperfluoro-3-oxaheptanesulphonyl fluoride
Vzorec:
C₆F₁₃IO₃S MW: 526.011 g/mol Bod varu: 87 °C (70 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD07778619 CAS číslo: 67990-76-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...