Tisk…

Ethyl perfluoro-2,5,8,11,14,17-hexamethyl-3,6,9,12,15,18-hexaoxaeicosanoate 95%

Dodavatel: Apollo Scientific
Varování

PC6826-5G PC6826-1G PC6826-25G
APOSPC6826-5GEA 11540 CZK
APOSPC6826-5G APOSPC6826-1G APOSPC6826-25G
Ethyl perfluoro-2,5,8,11,14,17-hexamethyl-3,6,9,12,15,18-hexaoxaeicosanoate 95%
Ethyl perfluoro-2,5,8,11,14,17-hexamethyl-3,6,9,12,15,18-hexaoxaeicosanoate
Vzorec: C₂₃H₅F₄₁O₈
Storage Temperature: Ambient
MDL Number: MFCD13176613
CAS číslo: 121368-60-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR